siRNA Details
| Gene Symbol: Pafah1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008775.3 | NP_032801.2 | 5 | 497 | ||
| GenBank mRNA | AK012164.1 | - | 5 | 432 | ||
| AK153424.1 | - | 5 | 468 | |||
| BC002037.1 | - | 5 | 445 | 3486003 | ||
| BC056211.2 | - | 5 | 467 | 30012448 | ||
| U57747.1 | - | 5 | 429 | |||

Pafah1b2
Genomic Map