siRNA Details
| Gene Symbol: Pafah1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008775.3 | NP_032801.2 | 2, 3 | 257 | ||
| GenBank mRNA | AK012116.1 | - | 2, 3 | 171 | ||
| AK012164.1 | - | 2, 3 | 192 | |||
| AK153424.1 | - | 2, 3 | 228 | |||
| BB856076.1 | - | 2, 3 | 258 | |||
| BC002037.1 | - | 2, 3 | 205 | 3486003 | ||
| BC056211.2 | - | 2, 3 | 227 | 30012448 | ||
| U57747.1 | - | 2, 3 | 189 | |||

Pafah1b2
Genomic Map