siRNA Details
| Gene Symbol: Pah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008777.3 | NP_032803.2 | 8 | 1101 | ||
| GenBank mRNA | AK145839.1 | - | 8 | 966 | ||
| AK149519.1 | - | 8 | 967 | |||
| BC013458.1 | - | 8 | 959 | 4223155 | ||
| X51942.1 | - | 8 | 926 | |||

Pah
Genomic Map