siRNA Details
| Gene Symbol: Pah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008777.3 | NP_032803.2 | 2 | 295 | ||
| GenBank mRNA | AK145839.1 | - | 2 | 161 | ||
| AK149519.1 | - | 2 | 161 | |||
| BC013458.1 | - | 2 | 153 | 4223155 | ||
| BY074411.1 | - | 2 | 300 | |||
| X51942.1 | - | 2 | 120 | |||

Pah
Genomic Map