siRNA Details
| Gene Symbol: Reg3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011036.1 | NP_035166.1 | 6 | 512 | ||
| GenBank mRNA | BC027525.1 | - | 5 | 495 | 3471932 | |
| D13509.1 | - | 485 | ||||
| D63359.1 | - | 6 | 512 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Reg3b| Gene Symbol: Reg3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011036.1 | NP_035166.1 | 6 | 512 | ||
| GenBank mRNA | BC027525.1 | - | 5 | 495 | 3471932 | |
| D13509.1 | - | 485 | ||||
| D63359.1 | - | 6 | 512 | |||