siRNA Details
| Gene Symbol: Pappa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021362.1 | NP_067337.1 | 7 | 2755 | ||
| GenBank mRNA | AF258461.1 | - | 6 | 1616 | ||
| AF439513.1 | - | 7 | 2150 | |||
| AF439514.1 | - | 8 | 2237 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pappa| Gene Symbol: Pappa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021362.1 | NP_067337.1 | 7 | 2755 | ||
| GenBank mRNA | AF258461.1 | - | 6 | 1616 | ||
| AF439513.1 | - | 7 | 2150 | |||
| AF439514.1 | - | 8 | 2237 | |||