siRNA Details
Gene Symbol: Pappa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021362.1 | NP_067337.1 | 2 | 1741 | ||
GenBank mRNA | AF258461.1 | - | 1 | 602 | ||
AF439513.1 | - | 2 | 1136 | |||
AF439514.1 | - | 2 | 1136 |
