siRNA Details
| Gene Symbol: Pax6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244198.1 | NP_001231127.1 | 6 | 635 | ||
| NM_001244200.1 | NP_001231129.1 | 7 | 847 | |||
| NM_001244201.1 | NP_001231130.1 | 5 | 593 | |||
| NM_001244202.1 | NP_001231131.1 | 5 | 1149 | |||
| NM_013627.5 | NP_038655.1 | 6 | 1191 | |||
| GenBank mRNA | AF443223.1 | - | 2 | 308 | ||
| AJ292077.1 | - | 5 | 452 | |||
| AJ307468.1 | - | 5 | 459 | |||
| AK028059.1 | - | 5 | 594 | |||
| AK032616.1 | - | 6 | 636 | |||
| AK045805.1 | - | 5 | 1150 | |||
| BC011272.1 | - | 6 | 879 | 4008490 | ||
| BC036957.1 | - | 5 | 589 | 4504106 | ||
| M77842.1 | - | 1 | 178 | |||
| X63963.1 | - | 6 | 512 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_013001, XM_006234634, XM_008762060, XM_006234633, XM_006234636, and others.

Pax6
Genomic Map