siRNA Details
| Gene Symbol: Pax8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011040.4 | NP_035170.1 | 6 | 735 | ||
| GenBank mRNA | AK050717.1 | - | 6 | 711 | ||
| AK052610.1 | - | 6 | 711 | |||
| BC020526.1 | - | 6 | 702 | 4239835 | ||
| X57487.1 | - | 6 | 692 | |||

Pax8
Genomic Map