siRNA Details
| Gene Symbol: Pcnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282992.1 | NP_001269921.1 | 22 | 4400 | ||
| NM_008787.3 | NP_032813.3 | 22 | 4400 | |||
| GenBank mRNA | AB207233.1 | - | 22 | 4274 | ||
| AB207234.1 | - | 10 | 1695 | |||
| AK122275.2 | - | 7 | 1147 | |||

Pcnt
Genomic Map