siRNA Details
| Gene Symbol: Pcsk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013628.2 | NP_038656.1 | 4 | 588 | ||
| GenBank mRNA | AK043742.1 | - | 4 | 660 | ||
| BC108982.1 | - | 4 | 566 | 40047652 | ||
| BC108983.1 | - | 4 | 566 | 40047655 | ||
| M58589.1 | - | 4 | 588 | |||
| M69196.1 | - | 4 | 694 | |||
| X57088.1 | - | 4 | 700 | |||

Pcsk1
Genomic Map