siRNA Details
| Gene Symbol: Pcsk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163144.1 | NP_001156616.1 | 5 | 1063 | ||
| NM_001190483.1 | NP_001177412.1 | 5 | 1063 | |||
| GenBank mRNA | AK086210.1 | - | 5 | 1050 | ||
| AK087337.1 | - | 5 | 1064 | |||
| AK143998.1 | - | 5 | 1064 | |||
| BC013068.1 | - | 5 | 683 | |||
| D12619.1 | - | 5 | 652 | |||
| L14932.1 | - | 5 | 613 | |||

Pcsk5
Genomic Map