siRNA Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162946.1 | NP_001156418.1 | 18, 19 | 3046 | ||
| NM_008797.3 | NP_032823.2 | 17, 18 | 3023 | |||
| GenBank mRNA | AK077665.1 | - | 9, 10 | 1851 | ||
| AK128927.1 | - | 2820 | ||||
| AK148328.1 | - | 18, 19 | 2993 | |||
| AK170596.1 | - | 18, 19 | 3006 | |||
| AK170871.1 | - | 18, 19 | 2997 | |||
| AK172320.1 | - | 18, 19 | 2974 | |||
| BC055030.1 | - | 18, 19 | 2981 | 5053475 | ||
| L09192.1 | - | 17, 18 | 2999 | |||
| M97957.1 | - | 12, 13 | 2171 | |||

Pcx
Genomic Map