siRNA Details
| Gene Symbol: Pde1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009978.1 | NP_001009978.1 | 3, 4 | 306 | ||
| NM_001009979.1 | NP_001009979.1 | 3, 4 | 306 | |||
| NM_001159582.1 | NP_001153054.1 | 5, 6 | 808 | |||
| NM_016744.4 | NP_058024.2 | 4, 5 | 603 | |||
| GenBank mRNA | AF023529.1 | - | 4, 5 | 523 | ||
| AF159298.1 | - | 3, 4 | 306 | |||
| AK014810.1 | - | 3, 4 | 305 | |||
| AK043647.1 | - | 4, 5 | 604 | |||
| AK050004.1 | - | 5, 6 | 669 | |||
| AY845863.1 | - | 4, 5 | 355 | |||
| AY845864.1 | - | 3, 4 | 306 | |||
| BC090628.1 | - | 4, 5 | 571 | 6842675 | ||
| U56649.1 | - | 5, 6 | 539 | |||

Pde1a
Genomic Map