siRNA Details
| Gene Symbol: Pdgfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083316.1 | NP_001076785.1 | 9 | 1534 | ||
| NM_011058.2 | NP_035188.2 | 9 | 1517 | |||
| GenBank mRNA | AK035501.1 | - | 9 | 1474 | ||
| AK081664.1 | - | 9 | 1516 | |||
| AK136490.1 | - | 9 | 1503 | |||
| AK139094.1 | - | 8 | 1749 | |||
| AK145097.1 | - | 9 | 2118 | |||
| AK147267.1 | - | 9 | 1466 | |||
| AK163952.1 | - | 9 | 1581 | |||
| BC020405.1 | - | 9 | 1448 | |||
| BC037467.1 | - | 9 | 1456 | |||
| BC053036.1 | - | 9 | 1534 | 5704645 | ||
| M84607.1 | - | 9 | 1527 | |||

Pdgfra
Genomic Map