siRNA Details
| Gene Symbol: Pdha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008810.2 | NP_032836.1 | 2 | 112 | ||
| GenBank mRNA | AK145718.1 | - | 2 | 190 | ||
| AK148459.1 | - | 2 | 133 | |||
| AK167987.1 | - | 2 | 138 | |||
| AK168152.1 | - | 2 | 162 | |||
| AK169289.1 | - | 2 | 187 | |||
| AK212590.1 | - | 1 | 33 | |||
| BC007142.1 | - | 2 | 112 | 3600605 | ||
| M76727.1 | - | 2 | 199 | |||

Pdha1
Genomic Map