siRNA Details
Gene Symbol: Padi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011059.2 | NP_035189.1 | 8 | 968 | ||
GenBank mRNA | AK037182.1 | - | 8 | 969 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Padi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011059.2 | NP_035189.1 | 8 | 968 | ||
GenBank mRNA | AK037182.1 | - | 8 | 969 |