siRNA Details
| Gene Symbol: Padi4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011061.2 | NP_035191.2 | 9, 10 | 1067 | ||
| GenBank mRNA | AB013850.1 | - | 9, 10 | 1060 | ||
| AK084904.1 | - | 3, 4 | 2440 | |||
| AK137625.1 | - | 9, 10 | 1069 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Padi4| Gene Symbol: Padi4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011061.2 | NP_035191.2 | 9, 10 | 1067 | ||
| GenBank mRNA | AB013850.1 | - | 9, 10 | 1060 | ||
| AK084904.1 | - | 3, 4 | 2440 | |||
| AK137625.1 | - | 9, 10 | 1069 | |||