siRNA Details
| Gene Symbol: Pdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133667.2 | NP_598428.2 | 4 | 614 | ||
| GenBank mRNA | AF267660.1 | - | 4 | 575 | ||
| AK002647.1 | - | 4 | 558 | |||
| BC021764.1 | - | 4 | 568 | 3488813 | ||
| CX223857.1 | - | 4 | 615 | |||

Pdk2
Genomic Map