siRNA Details
| Gene Symbol: Enpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF339910.1 | - | 20 | 1980 | ||
| AK029600.1 | - | 20 | 1889 | |||
| AK036952.1 | - | 20 | 1986 | |||
| AK041894.1 | - | 15 | 1350 | |||
| AK088857.1 | - | 20 | 1984 | |||
| AK145233.1 | - | 20 | 1980 | |||
| AK164619.1 | - | 20 | 1959 | |||
| BC050011.1 | - | 20 | 2131 | |||
| J02700.2 | - | 20 | 2088 | |||

Enpp1
Genomic Map