siRNA Details
| Gene Symbol: Enpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF339910.1 | - | 9 | 874 | ||
| AK029600.1 | - | 9 | 786 | |||
| AK035402.1 | - | 3 | 3016 | |||
| AK036952.1 | - | 9 | 883 | |||
| AK041894.1 | - | 4 | 247 | |||
| AK088857.1 | - | 9 | 881 | |||
| AK145233.1 | - | 9 | 877 | |||
| AK164619.1 | - | 9 | 882 | |||
| BC050011.1 | - | 9 | 1054 | |||
| J02700.2 | - | 9 | 985 | |||

Enpp1
Genomic Map