siRNA Details
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080773.2 | NP_001074242.1 | 12, 13 | 1552 | ||
NM_001286662.1 | NP_001273591.1 | 12, 13 | 1470 | |||
NM_011062.4 | NP_035192.2 | 12, 13 | 1552 | |||
GenBank mRNA | AF079535.1 | - | 12, 13 | 1509 | ||
AF086625.1 | - | 12, 13 | 1518 | |||
AF126294.1 | - | 12, 13 | 1493 | |||
AK147149.1 | - | 12, 13 | 1544 | |||
AK147835.1 | - | 12, 13 | 1527 | |||
AK149162.1 | - | 12, 13 | 1486 | |||
AK162677.1 | - | 12, 13 | 1512 | |||
AK165564.1 | - | 12, 13 | 1490 | |||
AY062008.1 | - | 12, 13 | 1448 | |||
BC146001.1 | - | 12, 13 | 1489 | 40131304 | ||
BC146003.1 | - | 12, 13 | 1489 | 40131306 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031081, XM_006246036, Y15748.
Panther Classification:
Gene Ontology Categories:
