siRNA Details
| Gene Symbol: Penk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002927.2 | NP_001002927.1 | 2 | 1051 | ||
| GenBank mRNA | AK135111.1 | - | 2 | 793 | ||
| AK161272.1 | - | 3 | 907 | |||
| BC049766.1 | - | 2 | 1051 | 6774387 | ||
| M13227.1 | - | 2 | 665 | |||
| M55181.1 | - | 2 | 1048 | |||

Penk
Genomic Map