siRNA Details
| Gene Symbol: Pex7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161825.1 | NP_001155297.1 | 9 | 926 | ||
| NM_008822.2 | NP_032848.1 | 10 | 1004 | |||
| GenBank mRNA | AK004687.1 | - | 10 | 989 | ||
| AK160195.1 | - | 10 | 968 | |||
| BC132213.1 | - | 10 | 977 | 40130490 | ||
| BC132503.1 | - | 10 | 977 | 40130780 | ||
| BC145300.1 | - | 9 | 899 | 9053854 | ||
| U69171.1 | - | 10 | 995 | |||

Pex7
Genomic Map