siRNA Details
| Gene Symbol: Pex7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161825.1 | NP_001155297.1 | 2 | 263 | ||
| NM_008822.2 | NP_032848.1 | 2 | 263 | |||
| GenBank mRNA | AK004687.1 | - | 2 | 248 | ||
| AK053868.1 | - | 1 | 1798 | |||
| AK160195.1 | - | 2 | 226 | |||
| BC132213.1 | - | 2 | 236 | 40130490 | ||
| BC132503.1 | - | 2 | 236 | 40130780 | ||
| BC145300.1 | - | 2 | 236 | 9053854 | ||
| DV660555.1 | - | 2 | 263 | |||
| U69171.1 | - | 2 | 254 | |||

Pex7
Genomic Map