siRNA Details
| Gene Symbol: Pfkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008826.4 | NP_032852.2 | 20 | 2134 | ||
| GenBank mRNA | AK030511.1 | - | 20 | 2089 | ||
| AK030892.1 | - | 13 | 1233 | |||
| AK036318.1 | - | 20 | 2067 | |||
| AK081313.1 | - | 20 | 2085 | |||
| AK159328.1 | - | 20 | 2079 | |||
| AK168660.1 | - | 20 | 2090 | |||
| AK216193.1 | - | 4 | 243 | |||
| BC020097.1 | - | 20 | 2088 | 4014395 | ||
| BC029336.1 | - | 20 | 2077 | |||
| J03928.1 | - | 20 | 2063 | |||

Pfkl
Genomic Map