siRNA Details
| Gene Symbol: Pgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008829.2 | NP_032855.2 | 2, 3 | 2407 | ||
| GenBank mRNA | AK036862.1 | - | 2, 3 | 1822 | ||
| AK054117.1 | - | 2, 3 | 1088 | |||
| AK142629.1 | - | 3, 4 | 729 | |||
| BC145807.1 | - | 2, 3 | 1812 | 40131110 | ||
| M68915.1 | - | 2, 3 | 1770 | |||

Pgr
Genomic Map