siRNA Details
| Gene Symbol: Pgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008829.2 | NP_032855.2 | 7 | 3149 | ||
| GenBank mRNA | AK054117.1 | - | 7 | 1830 | ||
| BC145807.1 | - | 7 | 2554 | 40131110 | ||
| M68915.1 | - | 7 | 2512 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgr| Gene Symbol: Pgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008829.2 | NP_032855.2 | 7 | 3149 | ||
| GenBank mRNA | AK054117.1 | - | 7 | 1830 | ||
| BC145807.1 | - | 7 | 2554 | 40131110 | ||
| M68915.1 | - | 7 | 2512 | |||