siRNA Details
| Gene Symbol: Phka1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008832.2 | NP_032858.2 | 25 | 3089 | ||
| NM_173021.3 | NP_766609.2 | 24 | 2912 | |||
| GenBank mRNA | AK036440.1 | - | 24 | 2865 | ||
| AK137497.1 | - | 24 | 2851 | |||
| X74616.1 | - | 25 | 2985 | |||

Phka1
Genomic Map