siRNA Details
Gene Symbol: Pik3c2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011083.2 | NP_035213.2 | 2 | 962 | ||
GenBank mRNA | AK049609.1 | - | 2 | 952 | ||
AK140908.1 | - | 3 | 1042 | |||
AK142284.1 | - | 3 | 991 | |||
BC096553.1 | - | 1 | 396 | |||
U52193.1 | - | 2 | 952 | |||
U55772.1 | - | 1 | 278 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
