siRNA Details
| Gene Symbol: Pik3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024955.2 | NP_001020126.1 | 6 | 982 | ||
| NM_001077495.2 | NP_001070963.1 | 12 | 2071 | |||
| GenBank mRNA | AK046259.1 | - | 6 | 985 | ||
| BC026146.1 | - | 12 | 2052 | 3979333 | ||
| BC051106.1 | - | 6 | 682 | 5031261 | ||
| CB519962.1 | - | 6 | 614 | |||
| M60651.1 | - | 12 | 2006 | |||
| U50413.1 | - | 12 | 2024 | |||

Pik3r1
Genomic Map