siRNA Details
| Gene Symbol: Pja1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083110.2 | NP_001076579.1 | 2 | 1337 | ||
| NM_001290555.1 | NP_001277484.1 | 2 | 1623 | |||
| NM_001290556.1 | NP_001277485.1 | 3 | 797 | |||
| NM_008853.3 | NP_032879.2 | 3 | 788 | |||
| GenBank mRNA | AF335250.2 | - | 2 | 1306 | ||
| AK005373.1 | - | 2 | 1244 | |||
| AK135382.1 | - | 2 | 1322 | |||
| AK145943.1 | - | 2 | 1580 | |||
| BC008148.1 | - | 1 | 611 | |||
| BC025975.1 | - | 2 | 1285 | 3983750 | ||
| BC037616.1 | - | 2 | 1260 | 5401000 | ||

Pja1
Genomic Map