siRNA Details
| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011101.3 | NP_035231.2 | 17 | 2189 | ||
| GenBank mRNA | AK147084.1 | - | 17 | 2190 | ||
| AK163950.1 | - | 7 | 877 | |||
| BC096493.1 | - | 17 | 1983 | 30742571 | ||
| M25811.1 | - | 17 | 2223 | |||
| X52684.1 | - | 18 | 2130 | |||
| X52685.1 | - | 18 | 2130 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001105713, XM_008768310, BC169007, X07286.

Prkca
Genomic Map