siRNA Details
| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011104.3 | NP_035234.1 | 5 | 1427 | ||
| GenBank mRNA | AF028009.1 | - | 5 | 628 | ||
| AF325507.1 | - | 5 | 619 | |||
| AK082438.1 | - | 5 | 1428 | |||
| AK085351.1 | - | 6 | 839 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkce| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011104.3 | NP_035234.1 | 5 | 1427 | ||
| GenBank mRNA | AF028009.1 | - | 5 | 628 | ||
| AF325507.1 | - | 5 | 619 | |||
| AK082438.1 | - | 5 | 1428 | |||
| AK085351.1 | - | 6 | 839 | |||