siRNA Details
| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011104.3 | NP_035234.1 | 7 | 1735 | ||
| GenBank mRNA | AF028009.1 | - | 7 | 936 | ||
| AK082438.1 | - | 7 | 1736 | |||
| AK085351.1 | - | 8 | 1147 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkce| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011104.3 | NP_035234.1 | 7 | 1735 | ||
| GenBank mRNA | AF028009.1 | - | 7 | 936 | ||
| AK082438.1 | - | 7 | 1736 | |||
| AK085351.1 | - | 8 | 1147 | |||