siRNA Details
| Gene Symbol: Prkcq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008859.2 | NP_032885.1 | 10 | 1001 | ||
| GenBank mRNA | AB062122.1 | - | 3 | 399 | ||
| AK037664.1 | - | 10 | 1073 | |||
| AK085546.1 | - | 10 | 1271 | |||
| BC138552.1 | - | 9 | 932 | 8861572 | ||
| BC145769.1 | - | 9 | 932 | 40131072 | ||
| D11091.1 | - | 10 | 1001 | |||

Prkcq
Genomic Map