siRNA Details
| Gene Symbol: Prkcq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008859.2 | NP_032885.1 | 9 | 967 | ||
| GenBank mRNA | AB062122.1 | - | 2 | 365 | ||
| AK037664.1 | - | 9 | 1039 | |||
| AK085546.1 | - | 9 | 1237 | |||
| BC138552.1 | - | 8 | 898 | 8861572 | ||
| BC145769.1 | - | 8 | 898 | 40131072 | ||
| D11091.1 | - | 9 | 967 | |||

Prkcq
Genomic Map