siRNA Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039079.2 | NP_001034168.1 | 5 | 671 | ||
| NM_008860.3 | NP_032886.2 | 8 | 871 | |||
| GenBank mRNA | AB110830.1 | - | 5 | 631 | ||
| AK042562.1 | - | 5 | 629 | |||
| AK049959.1 | - | 5 | 655 | |||
| AK147300.1 | - | 5 | 489 | |||
| BC072590.1 | - | 5 | 671 | |||
| BC139761.1 | - | 8 | 688 | 40109623 | ||
| M94632.1 | - | 8 | 688 | |||

Prkcz
Genomic Map