siRNA Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039079.2 | NP_001034168.1 | 12 | 1475 | ||
| NM_008860.3 | NP_032886.2 | 15 | 1675 | |||
| GenBank mRNA | AB110830.1 | - | 12 | 1435 | ||
| AK042562.1 | - | 12 | 1433 | |||
| AK049959.1 | - | 12 | 1459 | |||
| BC057694.1 | - | 7 | 642 | |||
| BC072590.1 | - | 12 | 1475 | |||
| BC139761.1 | - | 15 | 1492 | 40109623 | ||
| M94632.1 | - | 15 | 1492 | |||

Prkcz
Genomic Map