siRNA Details
| Gene Symbol: Pkdrej | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011105.2 | NP_035235.2 | 1 | 5947 | ||
| GenBank mRNA | AK029344.1 | - | 1 | 166 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pkdrej| Gene Symbol: Pkdrej | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011105.2 | NP_035235.2 | 1 | 5947 | ||
| GenBank mRNA | AK029344.1 | - | 1 | 166 | ||