siRNA Details
| Gene Symbol: Pknox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016670.3 | NP_057879.2 | 4 | 368 | ||
| GenBank mRNA | AF061270.1 | - | 4 | 340 | ||
| AK131788.1 | - | 3 | 397 | |||
| AK134000.1 | - | 5 | 525 | |||
| BC052701.1 | - | 3 | 263 | 5721441 | ||

Pknox1
Genomic Map