siRNA Details
| Gene Symbol: Pkp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019645.2 | NP_062619.1 | 10 | 1709 | ||
| GenBank mRNA | AK036856.1 | - | 10 | 1894 | ||
| AK079429.1 | - | 10 | 1894 | |||
| BC139041.1 | - | 10 | 1804 | 8862063 | ||
| BC139042.1 | - | 10 | 1805 | 8862064 | ||
| Y07941.1 | - | 10 | 1709 | |||

Pkp1
Genomic Map