siRNA Details
| Gene Symbol: Pa2g4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011119.3 | NP_035249.1 | 8, 9 | 943 | ||
| GenBank mRNA | AK002685.1 | - | 8, 9 | 1020 | ||
| AK076286.1 | - | 8, 9 | 948 | |||
| AK144645.1 | - | 8, 9 | 997 | |||
| AK166936.1 | - | 8, 9 | 952 | |||
| AK168582.1 | - | 8, 9 | 945 | |||
| BC003777.1 | - | 8, 9 | 933 | |||
| BC005593.1 | - | 8, 9 | 933 | |||
| BC028926.1 | - | 8, 9 | 926 | |||
| BC037623.1 | - | 8, 9 | 963 | |||
| BC046532.1 | - | 8, 9 | 897 | 5358060 | ||
| BC086788.1 | - | 6, 7 | 3149 | |||
| U43918.1 | - | 8, 9 | 1102 | |||
| X84789.1 | - | 8, 9 | 1008 | |||

Pa2g4
Genomic Map