siRNA Details
| Gene Symbol: Pln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141927.1 | NP_001135399.1 | 3 | 1382 | ||
| NM_023129.5 | NP_075618.1 | 2 | 1255 | |||
| GenBank mRNA | AF214653.1 | - | 1 | 496 | ||
| AK002622.1 | - | 2 | 1235 | |||
| AK040718.1 | - | 2 | 1225 | |||
| AK142708.1 | - | 2 | 1210 | |||

Pln
Genomic Map