siRNA Details
| Gene Symbol: Plod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011122.3 | NP_035252.1 | 13 | 1448 | ||
| GenBank mRNA | AK137480.1 | - | 13 | 1399 | ||
| BC006599.1 | - | 13 | 1373 | 3965301 | ||
| BC010268.1 | - | 5 | 393 | 3589182 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plod1| Gene Symbol: Plod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011122.3 | NP_035252.1 | 13 | 1448 | ||
| GenBank mRNA | AK137480.1 | - | 13 | 1399 | ||
| BC006599.1 | - | 13 | 1373 | 3965301 | ||
| BC010268.1 | - | 5 | 393 | 3589182 | ||