siRNA Details
| Gene Symbol: Plscr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195084.1 | NP_001182013.1 | 4 | 716 | ||
| NM_008880.3 | NP_032906.2 | 3 | 711 | |||
| GenBank mRNA | AF015790.1 | - | 4 | 482 | ||
| AK002410.1 | - | 3 | 598 | |||
| BC002012.1 | - | 4 | 582 | 3490475 | ||
| BC011299.1 | - | 3 | 593 | 3595010 | ||

Plscr2
Genomic Map