siRNA Details
| Gene Symbol: Pmp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076005.1 | - | 5 | 1412 | ||
| AK171745.1 | - | 5 | 1409 | |||
| BC010765.1 | - | 5 | 1435 | 4225316 | ||
| M32240.1 | - | 5 | 1462 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pmp22| Gene Symbol: Pmp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076005.1 | - | 5 | 1412 | ||
| AK171745.1 | - | 5 | 1409 | |||
| BC010765.1 | - | 5 | 1435 | 4225316 | ||
| M32240.1 | - | 5 | 1462 | |||