siRNA Details
| Gene Symbol: Phox2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008888.3 | NP_032914.1 | 1, 2 | 431 | ||
| GenBank mRNA | AK052579.2 | - | 1, 2 | 362 | ||
| BC079610.1 | - | 1, 2 | 431 | 30360139 | ||
| Y14493.1 | - | 1, 2 | 464 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phox2b| Gene Symbol: Phox2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008888.3 | NP_032914.1 | 1, 2 | 431 | ||
| GenBank mRNA | AK052579.2 | - | 1, 2 | 362 | ||
| BC079610.1 | - | 1, 2 | 431 | 30360139 | ||
| Y14493.1 | - | 1, 2 | 464 | |||