siRNA Details
| Gene Symbol: Phox2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008888.3 | NP_032914.1 | 2 | 465 | ||
| GenBank mRNA | AK052579.2 | - | 2 | 396 | ||
| BC079610.1 | - | 2 | 465 | 30360139 | ||
| Y14493.1 | - | 2 | 498 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phox2b| Gene Symbol: Phox2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008888.3 | NP_032914.1 | 2 | 465 | ||
| GenBank mRNA | AK052579.2 | - | 2 | 396 | ||
| BC079610.1 | - | 2 | 465 | 30360139 | ||
| Y14493.1 | - | 2 | 498 | |||