siRNA Details
| Gene Symbol: Pnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008891.2 | NP_032917.2 | 2, 3 | 272 | ||
| GenBank mRNA | AK009055.1 | - | 2, 3 | 227 | ||
| AK028219.1 | - | 2, 3 | 213 | |||
| AK030978.1 | - | 2, 3 | 201 | |||
| AK050669.1 | - | 2, 3 | 218 | |||
| AK051640.1 | - | 2, 3 | 270 | |||
| AK157587.1 | - | 2, 3 | 216 | |||
| AK160616.1 | - | 2, 3 | 207 | |||
| AK161109.1 | - | 2, 3 | 212 | |||
| BC132571.1 | - | 2, 3 | 206 | 40130848 | ||
| BC138014.1 | - | 2, 3 | 206 | 8861033 | ||
| BY775218.1 | - | 2, 3 | 273 | |||
| Y08701.1 | - | 2, 3 | 206 | |||

Pnn
Genomic Map